accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122775-35-3

IUPAC Name: (3,4-dimethoxyphenyl)boronic acid
Synonyms: 3,4-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: RCVDPBFUMYUKPB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)B(O)O
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (1)
Quantity 
  • (2)
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (7)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)