accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12279-09-3

IUPAC Name: bis(λ¹-rhodium(1+)) tetrakis((Z)-cyclooctene) dichloride
Synonyms: bis(λ¹-rhodium(1+)) tetrakis((Z)-cyclooctene) dichloride
Molecular Weight (g/mol): 717.51
Molecular Formula: C32H56Cl2Rh2
InChi Key: GQPAPAIPOLEZHT-XFCUKONHSA-L
SMILES: [Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)