accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1228776-03-1

IUPAC Name: 6-[amino(nitroso)methylidene]-1,6-dihydropyridin-3-amine
Synonyms: 6-[amino(nitroso)methylidene]-1H-pyridin-3-amine
Molecular Weight (g/mol): 152.16
Molecular Formula: C6H8N4O
InChi Key: HDHDMYYDFKTNEQ-UHFFFAOYSA-N
SMILES: NC(N=O)=C1NC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results