accessibility menu, dialog, popup

UserName

Viewing profile as user:
I

CAS RN 1228778-59-3

IUPAC Name: 2-iodotriphenylene
Synonyms: 2-iodotriphenylene
Molecular Weight (g/mol): 354.19
Molecular Formula: C18H11I
InChi Key: ZMZNYRZSJDYTIO-UHFFFAOYSA-N
SMILES: IC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results