CAS RN 122892-31-3
CAS RN 122892-31-3
IUPAC Name:
hydrogen N-({4-[2-(dimethylamino)ethoxy]phenyl}methyl)-3,4-dimethoxybenzamide chloride
Synonyms:
hydrogen itopride chloride
Molecular Weight (g/mol):
394.9
Molecular Formula:
C20H27ClN2O4
InChi Key:
ZTOUXLLIPWWHSR-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC1=CC=C(C=C1OC)C(=O)NCC1=CC=C(OCCN(C)C)C=C1
1
–
1
of
1
results
1
Itopride Hydrochloride 98.0+%, TCI America™
CAS: 122892-31-3 Molecular Formula: C20H27ClN2O4 Molecular Weight (g/mol): 394.896 MDL Number: MFCD00881710 InChI Key: ZTOUXLLIPWWHSR-UHFFFAOYSA-N Synonym: N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4-dimethoxybenzamide Hydrochloride PubChem CID: 129791 IUPAC Name: N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxybenzamide;hydrochloride SMILES: CN(C)CCOC1=CC=C(C=C1)CNC(=O)C2=CC(=C(C=C2)OC)OC.Cl