accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 122918-25-6

IUPAC Name: 6-bromopyridine-2-carbonitrile
Synonyms: 6-bromopyridine-2-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: HNEBPTAKURBYRM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)C#N
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)