accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1229603-71-7

IUPAC Name: 2,7-dibromo-9,9-difluoro-9H-fluorene
Synonyms: 2,7-dibromo-9,9-difluorofluorene
Molecular Weight (g/mol): 360
Molecular Formula: C13H6Br2F2
InChi Key: ITRVPZLDBPKHTI-UHFFFAOYSA-N
SMILES: FC1(F)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results