accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CN

CAS RN 123-03-5

IUPAC Name: 1-hexadecylpyridin-1-ium chloride
Synonyms: cetylpyridinium chloride cetylpyridinium chloride anhydrous
Molecular Weight (g/mol): 339.99
Molecular Formula: C21H38ClN
InChi Key: YMKDRGPMQRFJGP-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)