accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOCH2

CAS RN 123-34-2

IUPAC Name: 3-(prop-2-en-1-yloxy)propane-1,2-diol
Synonyms: glycerin monoallyl ether
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: PAKCOSURAUIXFG-UHFFFAOYNA-N
SMILES: OCC(O)COCC=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results