accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-35-3

IUPAC Name: 7-methyl-3-methylideneocta-1,6-diene
Synonyms: α-myrcene β-myrcene myrcene (natural)
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: UAHWPYUMFXYFJY-UHFFFAOYSA-N
SMILES: CC(C)=CCCC(=C)C=C
Percent Purity 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)