accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(E)

CAS RN 123-73-9

IUPAC Name: (2E)-but-2-enal
Synonyms: crotonaldehyde 1-formylpropene crotylaldehyde methylpropenal β-methylacrolein aldehyde crotonique propylene aldehyde
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: MLUCVPSAIODCQM-NSCUHMNNSA-N
SMILES: C\C=C\C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)