accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 123-76-2

IUPAC Name: 4-oxopentanoic acid
Synonyms: levulinic acid acidum laevulinicum g-ketovalerate 4-oxopentansaeure levulic acid g-ketovaleric acid levulinsaeure usaf CZ-1 laevulic acid LEVA
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: JOOXCMJARBKPKM-UHFFFAOYSA-N
SMILES: CC(=O)CCC(O)=O
Boiling Point 
  • (2)
  • (3)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (2)
  • (5)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (7)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)