accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123-81-9

IUPAC Name: 2-[(2-sulfanylacetyl)oxy]ethyl 2-sulfanylacetate
Synonyms: 2-[(2-sulfanylacetyl)oxy]ethyl 2-sulfanylacetate
Molecular Weight (g/mol): 210.26
Molecular Formula: C6H10O4S2
InChi Key: PSYGHMBJXWRQFD-UHFFFAOYSA-N
SMILES: SCC(=O)OCCOC(=O)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results