accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOClO

CAS RN 123-98-8

IUPAC Name: nonanedioyl dichloride
Synonyms: nonanedioyl dichloride
Molecular Weight (g/mol): 225.11
Molecular Formula: C9H14Cl2O2
InChi Key: HGEVGSTXQGZPCL-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)