accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123071-42-1

IUPAC Name: N1,N3-diphenylpropane-1,3-diimine hydrochloride
Synonyms: N1,N3-diphenylpropane-1,3-diimine hydrochloride
Molecular Weight (g/mol): 258.75
Molecular Formula: C15H15ClN2
InChi Key: FLMLOZWXXPLHIE-UHFFFAOYSA-N
SMILES: Cl.C(C=NC1=CC=CC=C1)C=NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results