accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123266-59-1

IUPAC Name: 5,7-dibromo-2,3-dihydro-1-benzofuran
Synonyms: 5,7-dibromo-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 277.94
Molecular Formula: C8H6Br2O
InChi Key: NJQJUJHJJZMVNB-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C2OCCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results