accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1232692-92-0

CH3H3CSiNCH3CH3

CAS RN 1232692-92-0

IUPAC Name: bis(propan-2-yl)(pyridin-2-yl)silyl
Synonyms: diisopropyl(pyridin-2-yl)silyl
Molecular Weight (g/mol): 192.36
Molecular Formula: C11H18NSi
InChi Key: MARDZIUPIJQZBI-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)