accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1233026-65-7

IUPAC Name: 3-fluoro-2-(piperazin-1-yl)benzonitrile
Synonyms: 3-fluoro-2-(piperazin-1-yl)benzonitrile
Molecular Weight (g/mol): 205.24
Molecular Formula: C11H12FN3
InChi Key: BSCQTNZKNNTAGP-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C#N)=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results