accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123324-71-0

IUPAC Name: (4-tert-butylphenyl)boronic acid
Synonyms: 4-tert-butylphenylboronic acid
Molecular Weight (g/mol): 178.04
Molecular Formula: C10H15BO2
InChi Key: MNJYZNVROSZZQC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results