accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CSOOOH2ONa

CAS RN 123333-94-8

IUPAC Name: sodium 4-ethenylbenzene-1-sulfonate hydrate
Synonyms: sodium 4-ethenylbenzenesulfonate hydrate
Molecular Weight (g/mol): 224.21
Molecular Formula: C8H9NaO4S
InChi Key: AATHLPHPRXGBAI-UHFFFAOYSA-M
SMILES: O.[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)