CAS RN 123334-05-4
CAS RN 123334-05-4
IUPAC Name:
zinc(2+) bis(4-methylbenzene-1-sulfonate)
Synonyms:
zinc(2+) ditosylate
Molecular Weight (g/mol):
407.76
Molecular Formula:
C14H14O6S2Zn
InChi Key:
YISPIDBWTUCKKH-UHFFFAOYSA-L
SMILES:
[Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Zinc p-toluenesulfonate hydrate, Thermo Scientific™
CAS: 123334-05-4 Molecular Formula: C14H14O6S2Zn Molecular Weight (g/mol): 407.76 MDL Number: MFCD00151013 InChI Key: YISPIDBWTUCKKH-UHFFFAOYSA-L Synonym: zinc p-toluenesulfonate hydrate,acmc-20eh0l,p-toluenesulfonic acid zinc salt,zinc 2+ hydrate ditosylate,p-toluenesulfonic acid zinc salt hydrate,benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:? PubChem CID: 16212267 IUPAC Name: zinc(2+) bis(4-methylbenzene-1-sulfonate) SMILES: [Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O