accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaClIrClClClClCl

CAS RN 123334-23-6

IUPAC Name: trisodium hexachloroiridiumtriuide
Synonyms: trisodium hexachloroiridiumtriuide
Molecular Weight (g/mol): 473.89
Molecular Formula: Cl6IrNa3
InChi Key: GSONQQIOULBMRS-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Na+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)