accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123334-29-2

IUPAC Name: bis(4-hydroxypent-3-en-2-one) cobalt hydrate
Synonyms: bis(4-hydroxypent-3-en-2-one) cobalt hydrate
Molecular Weight (g/mol): 277.18
Molecular Formula: C10H18CoO5
InChi Key: JHWSVOFBMAXGJH-UHFFFAOYSA-N
SMILES: O.[Co].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results