accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH2NNOO

CAS RN 1233513-42-2

IUPAC Name: 4-methyl-3-nitrobenzene-1-carbothioamide
Synonyms: 4-methyl-3-nitrobenzenecarbothioamide
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: PGQMEJDSPAENNP-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results