accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNFNOO

CAS RN 1233952-28-7

IUPAC Name: 1-(2-fluoro-6-nitrophenyl)-4-methylpiperazine
Synonyms: 1-(2-fluoro-6-nitrophenyl)-4-methylpiperazine
Molecular Weight (g/mol): 239.25
Molecular Formula: C11H14FN3O2
InChi Key: WIAGMIDRHKDTGU-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C(C=CC=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results