accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNFNOO

CAS RN 1233958-36-5

IUPAC Name: tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 325.34
Molecular Formula: C15H20FN3O4
InChi Key: GWHUVEIGDXIWOY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=C(C=CC=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results