CAS RN 123409-01-8
CAS RN 123409-01-8
IUPAC Name:
naphthalene-1,3,6-trisulfonic acid hydrate trisodium
Synonyms:
naphthalene-1,3,6-trisulfonic acid hydrate trisodium
Molecular Weight (g/mol):
455.33
Molecular Formula:
C10H10Na3O10S3
InChi Key:
BMCNZRJPDRPEDI-UHFFFAOYSA-N
SMILES:
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
1
–
2
of
2
results
1
Naphthalene-1,3,6-trisulfonic Acid Trisodium Salt Hydrate
CAS: 123409-01-8 Molecular Formula: C10H10Na3O10S3 Molecular Weight (g/mol): 455.33 MDL Number: MFCD00149263 InChI Key: BMCNZRJPDRPEDI-UHFFFAOYSA-N Synonym: 1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate PubChem CID: 123134558 IUPAC Name: naphthalene-1,3,6-trisulfonic acid;sodium;hydrate SMILES: O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
2
Sigma Aldrich 1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.