CAS RN 123417-18-5
IUPAC Name:
(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[(9H-fluoren-9-yl)methoxy]-5-oxopentanoic acid
Synonyms:
(2S)-2-[(tert-butoxycarbonyl)amino]-5-(9H-fluoren-9-ylmethoxy)-5-oxopentanoic acid
Molecular Weight (g/mol):
425.48
Molecular Formula:
C24H27NO6
InChi Key:
RCQRXYWDDVULAP-FQEVSTJZSA-N
SMILES:
CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Boc-Glu OFm -OH, Advanced ChemTech
CAS: 123417-18-5 Molecular Formula: C24H27NO6 Molecular Weight (g/mol): 425.48 InChI Key: RCQRXYWDDVULAP-FQEVSTJZSA-N IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[(9H-fluoren-9-yl)methoxy]-5-oxopentanoic acid SMILES: CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O