accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3OP

CAS RN 1235-21-8

IUPAC Name: (2-oxopropyl)triphenylphosphanium chloride
Synonyms: (2-oxopropyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 354.81
Molecular Formula: C21H20ClOP
InChi Key: XAMZZEBAJZJERT-UHFFFAOYSA-M
SMILES: [Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)