CAS RN 1235005-44-3
IUPAC Name:
(2S)-3-(2,6-difluorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
(2S)-3-(2,6-difluorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
423.42
Molecular Formula:
C24H19F2NO4
InChi Key:
WNKYWXMLHBTJJW-QFIPXVFZSA-N
SMILES:
OC(=O)[C@H](CC1=C(F)C=CC=C1F)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Fmoc-Phe 2.6-DiF -OH, Advanced ChemTech
CAS: 1235005-44-3 Molecular Formula: C24H19F2NO4 Molecular Weight (g/mol): 423.42 MDL Number: MFCD07372009 InChI Key: WNKYWXMLHBTJJW-QFIPXVFZSA-N IUPAC Name: (2S)-3-(2,6-difluorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid SMILES: OC(=O)[C@H](CC1=C(F)C=CC=C1F)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2