accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3H3COH3CNSOOFSOOF

CAS RN 1235234-35-1

IUPAC Name: bis(fluorosulfonyl)azanide; ethyl(2-methoxyethyl)dimethylazanium
Synonyms: bis(fluorosulfonyl)azanide; ethyl(2-methoxyethyl)dimethylazanium
Molecular Weight (g/mol): 312.35
Molecular Formula: C7H18F2N2O5S2
InChi Key: JEXCPIWYTQMZON-UHFFFAOYSA-N
SMILES: CC[N+](C)(C)CCOC.FS(=O)(=O)[N-]S(F)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results