accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOH3CNSOOFSOOF

CAS RN 1235234-47-5

IUPAC Name: 1-(2-methoxyethyl)-1-methylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Synonyms: 1-(2-methoxyethyl)-1-methylpyrrolidin-1-ium; bis(fluorosulfonyl)azanide
Molecular Weight (g/mol): 324.36
Molecular Formula: C8H18F2N2O5S2
InChi Key: HBTKRUXLKWRXSK-UHFFFAOYSA-N
SMILES: FS(=O)(=O)[N-]S(F)(=O)=O.COCC[N+]1(C)CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results