accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123536-14-1

IUPAC Name: 1-azabicyclo[2.2.2]octan-3-amine dihydrochloride
Synonyms: 1-azabicyclo[2.2.2]octan-3-amine dihydrochloride
Molecular Weight (g/mol): 199.12
Molecular Formula: C7H16Cl2N2
InChi Key: STZHBULOYDCZET-UHFFFAOYNA-N
SMILES: Cl.Cl.NC1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)