accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFF

CAS RN 123577-99-1

IUPAC Name: 1-(3,5-difluorophenyl)ethan-1-one
Synonyms: 1-(3,5-difluorophenyl)ethanone
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: OXJLDNSPGPBDCP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)