accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1236349-67-9

IUPAC Name: 8-bromo-5H-pyrido[3,2-b]indole
Synonyms: 8-bromo-5H-pyrido[3,2-b]indole
Molecular Weight (g/mol): 247.1
Molecular Formula: C11H7BrN2
InChi Key: OXYFZHUGVBINOL-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC3=C2N=CC=C3)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results