accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123654-26-2

IUPAC Name: 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
Synonyms: 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
Molecular Weight (g/mol): 213.62
Molecular Formula: C9H8ClNO3
InChi Key: KRMUVKSAOVLXLF-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C(O)=O)C2=C1CCO2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results