accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2PHOO

CAS RN 123691-14-5

IUPAC Name: (3-aminopropyl)(cyclohexylmethyl)phosphinic acid
Synonyms: 3-aminopropyl(cyclohexylmethyl)phosphinic acid
Molecular Weight (g/mol): 219.27
Molecular Formula: C10H22NO2P
InChi Key: XOESDNIUAWGCLU-UHFFFAOYNA-N
SMILES: NCCCP(O)(=O)CC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results