accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123694-03-1

IUPAC Name: 4-(3,4-dihydroxyphenyl)but-3-en-2-one
Synonyms: 4-(3,4-dihydroxyphenyl)but-3-en-2-one
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: YIFZKRGUGKLILR-UHFFFAOYSA-N
SMILES: CC(=O)C=CC1=CC(O)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results