CAS RN 123707-26-6
IUPAC Name:
1,3-dimethoxy-1,3-dimethylurea
Synonyms:
1,3-dimethoxy-1,3-dimethylurea
Molecular Weight (g/mol):
148.16
Molecular Formula:
C5H12N2O3
InChi Key:
BXBKNWRSAAAPKG-UHFFFAOYSA-N
SMILES:
CON(C)C(=O)N(C)OC
1
–
1
of
1
results
1
1,3-Dimethoxy-1,3-dimethylurea 97.0+%, TCI America™
CAS: 123707-26-6 Molecular Formula: C5H12N2O3 Molecular Weight (g/mol): 148.16 MDL Number: MFCD22041645 InChI Key: BXBKNWRSAAAPKG-UHFFFAOYSA-N PubChem CID: 11159391 IUPAC Name: 1,3-dimethoxy-1,3-dimethylurea SMILES: CON(C)C(=O)N(C)OC