accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123770-62-7

IUPAC Name: ethyl 5-(hydroxymethyl)-1,2-oxazole-3-carboxylate
Synonyms: ethyl 5-(hydroxymethyl)-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol): 171.15
Molecular Formula: C7H9NO4
InChi Key: ZWVVVEYXDQMHGQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NOC(CO)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results