CAS RN 123863-97-8
IUPAC Name:
9,9-dihexyl-9H-fluorene
Synonyms:
9,9-dihexylfluorene
Molecular Weight (g/mol):
334.55
Molecular Formula:
C25H34
InChi Key:
LQQKFGSPUYTIRB-UHFFFAOYSA-N
SMILES:
CCCCCCC1(CCCCCC)C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
9,9-Dihexylfluorene 98.0+%, TCI America™
CAS: 123863-97-8 Molecular Formula: C25H34 Molecular Weight (g/mol): 334.55 MDL Number: MFCD14584648 InChI Key: LQQKFGSPUYTIRB-UHFFFAOYSA-N PubChem CID: 5128449 IUPAC Name: 9,9-dihexyl-9H-fluorene SMILES: CCCCCCC1(CCCCCC)C2=CC=CC=C2C2=CC=CC=C12