accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 123863-99-0

IUPAC Name: 9,9-dioctyl-9H-fluorene
Synonyms: 9,9-dioctylfluorene
Molecular Weight (g/mol): 390.66
Molecular Formula: C29H42
InChi Key: RXACYPFGPNTUNV-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results