accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ONOOCH3OH

CAS RN 1239468-48-4

IUPAC Name: methyl 3-[9-(dimethylamino)-N-hydroxynonanamido]propanoate
Synonyms: methyl 3-[9-(dimethylamino)-N-hydroxynonanamido]propanoate
Molecular Weight (g/mol): 302.42
Molecular Formula: C15H30N2O4
InChi Key: KDYRPQNFCURCQB-UHFFFAOYSA-N
SMILES: COC(=O)CCN(O)C(=O)CCCCCCCCN(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results