accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1239481-05-0

IUPAC Name: 2-[4-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[4-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 259.12
Molecular Formula: C13H18BN3O2
InChi Key: CARNXRHOIGMOQD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN=[N+]=[N-])C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results