accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-02-7

IUPAC Name: bis(prop-2-en-1-yl)amine
Synonyms: diallylamine
Molecular Weight (g/mol): 97.16
Molecular Formula: C6H11N
InChi Key: DYUWTXWIYMHBQS-UHFFFAOYSA-N
SMILES: C=CCNCC=C
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)