accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-10-7

IUPAC Name: methyl tetradecanoate
Synonyms: methyl myristate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: ZAZKJZBWRNNLDS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (7)
  • (6)
  • (2)
  • (1)