accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-21-0

IUPAC Name: bis(2-ethoxyethyl)amine
Synonyms: bis(2-ethoxyethyl)amine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: QDZOFZFDBDYWJX-UHFFFAOYSA-N
SMILES: CCOCCNCCOCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results