accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 124-25-4

IUPAC Name: tetradecanal
Synonyms: tetradecanal
Molecular Weight (g/mol): 212.38
Molecular Formula: C14H28O
InChi Key: UHUFTBALEZWWIH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results