accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124-30-1

IUPAC Name: octadecan-1-amine
Synonyms: octadecylamine hydrogenated tallowamine
Molecular Weight (g/mol): 269.52
Molecular Formula: C18H39N
InChi Key: REYJJPSVUYRZGE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCN
Boiling Point 
  • (2)
  • (6)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (6)
  • (7)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)