accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClBrBr

CAS RN 124-48-1

IUPAC Name: dibromo(chloro)methane
Synonyms: dibromochloromethane C-H-br2-cl chbr2cl ccris 938 CDBM
Molecular Weight (g/mol): 208.28
Molecular Formula: CHBr2Cl
InChi Key: GATVIKZLVQHOMN-UHFFFAOYSA-N
SMILES: ClC(Br)Br
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)